Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01345663_m1
          See other EPHX3 GE Assays ›
          Gene Symbol:
          Ephx3
          Gene Name:
          epoxide hydrolase 3
          Gene Aliases:
          2310063B19Rik, Abhd9
          Chromosome Location:
          Chr.17: 32183770 - 32189850 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01345663_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ephx3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033163.3 NP_001028335.1 4 - 5 821 69
          GenBank mRNA AK010021.1 - 4 - 5 810 69
          AK132483.1 - 4 - 5 822 69
          BC145797.1 - 4 - 5 886 40131100 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001835.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ephx3

          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          catalytic activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31144190 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31144187 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31144188 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31144189 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051825 AK132483.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051825 NM_001033163.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 894619 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 880849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 860532 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 10871 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123811 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051825 AK132483.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051825 NM_001033163.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31144180 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123811 - -
          Whole Mouse Genome G4122A A_51_P178114 AK132483.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P178114 BC145797.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P178114 NM_001033163.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P178114 AK010021.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123811 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31144187 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31144189 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31144190 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31144188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31144136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13261372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13415342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13494555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13261375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13561899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13575402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13471631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13415345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13308946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13141726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13352494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13581983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13308944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13261377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13366101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13560052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13141725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13141727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13081470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13324699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13261380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31144148 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10449811 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449812 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449810 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449808 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449813 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449814 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449809 - -
          Affymetrix HT MG-430 PM Array Plate 1430724_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10449807 - -
          Mouse Genome 430 2.0 Array 1430724_at - -
          Mouse Genome 430 2.0 Array 1430724_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71932_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71932_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700016897.mm.1 NM_001033163.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016897.mm.1 NM_001033163.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016897.mm.1 TC1700001835.mm.1 MTA TC
          Mouse Expression Set 430 1430724_at - -
          Mouse Expression Set 430 1430724_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430724_at - -
          Affymetrix HT Mouse Genome 430B Array 1430724_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 319500 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222751 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222751 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline