Search
Search

Akap8l| Gene Symbol: Akap8l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017476.2 | NP_059504.2 | 8 - 9 | 1127 | 128 | |
| GenBank mRNA | AK011325.1 | - | 8 - 9 | 1132 | 128 | |
| AK166526.1 | - | 9 - 10 | 1043 | 128 | ||
| BC046801.1 | - | 8 - 9 | 1116 | 6403966 | 128 | |
Set Membership: |
|