Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01346744_m1
          See other SMC2OS GE Assays ›
          Gene Symbol:
          Smc2os
          Gene Name:
          structural maintenance of chromosomes 2, opposite strand
          Gene Aliases:
          2700081L22Rik
          Chromosome Location:
          Chr.4: 52430284 - 52438964 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01346744_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smc2os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045175.1 1 - 2 306 72
          GenBank mRNA AK012547.1 - 1 - 2 142 72
          BE944567.1 - 1 - 2 72
          BX515870.1 - 1 - 2 306 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002657.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 433661 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855631 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94672 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455242 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 106925 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2235184 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05237057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22917546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22917545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22917543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22917542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22917534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22917533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22917547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22917537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22917539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22917532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22917540 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1441142_PM_at - -
          Mouse Genome 430 2.0 Array 1441142_PM_at - -
          Mouse Genome 430 2.0 Array 1441142_at - -
          Affymetrix Mouse Clariom S Array PSR0400021571.mm.1 NR_045175.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021571.mm.1 NR_045175.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021571.mm.1 TC0400002657.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021572.mm.1 NR_045175.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021572.mm.1 NR_045175.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021572.mm.1 TC0400002657.mm.1 MTA TC
          Mouse Expression Set 430 1441142_PM_at - -
          Mouse Expression Set 430 1441142_at - -
          Murine Genome U74v2 Set 136968_at - -
          Murine Genome U74v2 Set 136968_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441142_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441142_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 508395 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline