Search
Search

Vmn1r11| Gene Symbol: Vmn1r11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053233.2 | NP_444463.2 | 1 | 163 | 141 | |
| GenBank mRNA | BC104398.1 | - | 1 | 163 | 40050459 | 141 |
| BC104399.1 | - | 1 | 163 | 40050460 | 141 | |
Set Membership: |
|