Search
Search

Ubash3a| Gene Symbol: Ubash3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177823.4 | NP_808491.2 | 8 - 9 | 1209 | 97 | |
| GenBank mRNA | AK040572.1 | - | 8 - 9 | 1170 | 97 | |
| AK041690.1 | - | 8 - 9 | 1176 | 97 | ||
| AK049106.1 | - | 7 - 8 | 1109 | 97 | ||
| AK169696.1 | - | 8 - 9 | 1164 | 97 | ||
Set Membership: |
|