Search
Search

Cep44| Gene Symbol: Cep44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009951.1 | NP_001009951.1 | 5 - 6 | 584 | 64 | |
| GenBank mRNA | AK220415.1 | - | 4 - 5 | 540 | 64 | |
| BC088983.1 | - | 5 - 6 | 584 | 5700628 | 64 | |
Set Membership: |
|