Search
Search
| Gene Symbol: Ctrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033875.2 | NP_001029047.1 | 5 - 6 | 585 | 103 | |
| GenBank mRNA | AK007772.1 | - | 4 - 5 | 413 | 103 | |
| BC103715.1 | - | 5 - 6 | 583 | 6430373 | 103 | |
| BC115516.1 | - | 5 - 6 | 585 | 40101127 | 103 | |
| BC115517.1 | - | 5 - 6 | 585 | 40101128 | 103 | |
Set Membership: |
|