Search
Search

Fbxw17| Gene Symbol: Fbxw17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175401.3 | NP_780610.2 | 3 - 4 | 309 | 67 | |
| GenBank mRNA | AK135553.1 | - | 3 - 4 | 323 | 67 | |
| AK155157.1 | - | 3 - 4 | 408 | 67 | ||
| BC040428.1 | - | 3 - 4 | 309 | 3327345 | 67 | |
Set Membership: |
|