Search
Search

Spock3| Gene Symbol: Spock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252620.1 | NP_001239549.1 | 5 - 6 | 430 | 111 | |
| NM_001252621.1 | NP_001239550.1 | 4 - 5 | 421 | 111 | ||
| NM_023689.3 | NP_076178.1 | 5 - 6 | 544 | 111 | ||
| GenBank mRNA | AJ278998.1 | - | 5 - 6 | 438 | 111 | |
| AK013644.1 | - | 5 - 6 | 473 | 111 | ||
| AK049843.1 | - | 4 - 5 | 422 | 111 | ||
| AK220280.1 | - | 5 - 6 | 528 | 111 | ||
| BC017601.1 | - | 5 - 6 | 443 | 4502169 | 111 | |
| BC053334.1 | - | 5 - 6 | 415 | 6492113 | 111 | |
Set Membership: |
|