Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01349584_g1
          See other ARHGEF39 GE Assays ›
          Gene Symbol:
          Arhgef39
          Gene Name:
          Rho guanine nucleotide exchange factor (GEF) 39
          Gene Aliases:
          E130306D19Rik
          Chromosome Location:
          Chr.4: 43496139 - 43508769 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01349584_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arhgef39
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013377.2 NP_001013395.2 2 - 3 283 64
          GenBank mRNA AK153880.1 - 2 - 3 278 64
          BC080700.1 - 2 - 3 255 30472977 64
          BY009353.1 - 2 - 3 285 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002527.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell migration
          regulation of Rho protein signal transduction
          molecular_function
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22872022 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22872021 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133220 BC080700.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133220 AK087493.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133220 AK153880.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133220 NM_001013377.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266943 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623937 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133220 BC080700.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133220 AK087493.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133220 AK153880.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133220 NM_001013377.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872065 - -
          Whole Mouse Genome G4122A A_52_P27878 AK163287.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P27878 BC080700.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P27878 AK087493.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P27878 AK153880.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P27878 NM_001013377.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P143671 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03918941 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03950039 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03950038 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03764162 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03918943 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03918942 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03918944 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03950026 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03950031 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03950030 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22872022 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22872021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03722895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03366443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03808421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03918944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03797269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03764162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03722881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03808413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872038 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10512497 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512493 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512490 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512496 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512494 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512495 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512492 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512498 - -
          Affymetrix HT MG-430 PM Array Plate 1436725_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10512489 - -
          Mouse Genome 430 2.0 Array 1436725_at - -
          Mouse Genome 430 2.0 Array 1436725_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230098_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230098_at - -
          Affymetrix Mouse Clariom S Array PSR0400020549.mm.1 TC0400002527.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020549.mm.1 NM_001013377.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020549.mm.1 NM_001013377.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020548.mm.1 TC0400002527.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020548.mm.1 NM_001013377.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020548.mm.1 NM_001013377.2 Refseq
          Mouse Expression Set 430 1436725_at - -
          Mouse Expression Set 430 1436725_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436725_at - -
          Affymetrix HT Mouse Genome 430B Array 1436725_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 348732 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1230170 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2659314 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2659314 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2659317 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline