| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300015526.mm.1 |
NM_134071.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015526.mm.1 |
NM_134071.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015526.mm.1 |
TC1300002347.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300015527.mm.1 |
NM_134071.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015527.mm.1 |
NM_134071.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015527.mm.1 |
TC1300002347.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300015529.mm.1 |
NM_134071.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015529.mm.1 |
NM_134071.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300015529.mm.1 |
TC1300002347.mm.1 |
MTA TC |