Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01350213_m1
          See other OTUD3 GE Assays ›
          Gene Symbol:
          Otud3
          Gene Name:
          OTU domain containing 3
          Gene Aliases:
          3110030K17Rik
          Chromosome Location:
          Chr.4: 138891063 - 138913947 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01350213_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Otud3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028453.1 NP_082729.1 7 - 8 1129 57
          GenBank mRNA AK014098.1 - 7 - 8 1165 57

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          protein K11-linked deubiquitination
          protein K6-linked deubiquitination
          thiol-dependent ubiquitin-specific protease activity
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087923 NM_028453.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087923 AK014098.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 157486 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732329 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 258907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36356 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 340748 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145751 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911717 AK036136.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911717 AY149280.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911717 AY149281.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911717 BC019671.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911717 BC027078.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911717 BC076593.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911717 NM_175328.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 421368 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715831 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430665 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 115110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126888 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143973 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101570 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087923 NM_028453.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087923 AK014098.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101570 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143973 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01002679 - -
          Whole Mouse Genome G4122A A_51_P381397 NM_028453.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P381397 AK014098.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101570 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03278910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03849691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04030377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03467734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03443003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03786253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03995418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03878018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03849690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03825164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03878036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03878017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03849692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03848218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348646 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430046_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10517666 - -
          Mouse Genome 430 2.0 Array 1430046_PM_at - -
          Mouse Genome 430 2.0 Array 1430046_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73162_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73162_at - -
          Mouse Expression Set 430 1430046_at - -
          Mouse Expression Set 430 1430046_PM_at - -
          Murine Genome U74v2 Set 139514_PM_r_at - -
          Murine Genome U74v2 Set 139514_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1430046_at - -
          Affymetrix HT Mouse Genome 430B Array 1430046_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 585410 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2615134 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2615134 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline