Search
Search
| Gene Symbol: Ice1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144837.3 | NP_659086.2 | 16 - 17 | 6350 | 91 | |
| GenBank mRNA | AK129244.1 | - | 4 - 5 | 4787 | 91 | |
| AK134564.1 | - | 4 - 5 | 2793 | 91 | ||
| BC018507.1 | - | 3 - 4 | 445 | 91 | ||
| BC026613.1 | - | 4 - 5 | 876 | 91 | ||
Set Membership: |
|