Search
Search
| Gene Symbol: Tff3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011575.2 | NP_035705.1 | 1 - 2 | 114 | 64 | |
| GenBank mRNA | BC011042.1 | - | 1 - 2 | 109 | 4216708 | 64 |
| D38410.1 | - | 1 - 2 | 114 | 64 | ||
Set Membership: |
|