Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01351724_g1
          See other SERPINB6D GE Assays ›
          Gene Symbol:
          Serpinb6d
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade B, member 6d
          Gene Aliases:
          Gm11390, OTTMUSG00000000712, SPI3D
          Chromosome Location:
          Chr.13: 33661405 - 33671585 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01351724_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpinb6d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001076790.2 NP_001070258.1 6 - 7 959 112
          NM_001300892.1 NP_001287821.1 6 - 7 804 112
          GenBank mRNA AK136270.1 - 6 - 7 961 112
          BC147559.1 - 6 - 7 804 9087441 112

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          negative regulation of endopeptidase activity
          molecular_function
          serine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P602669 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447575 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193451 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 912213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 97888 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 160219 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P602669 - -
          Whole Mouse Genome G4122A A_52_P602669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28934781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10563553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10725542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10934698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10638237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10673451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10604091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10585117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10638254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10854836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10638250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10604106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28934780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10604079 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404494 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404492 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404495 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404488 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404493 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404490 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404489 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404487 - -
          Affymetrix Mouse Clariom S Array PSR1300002263.mm.1 NM_001076790.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002263.mm.1 NM_001076790.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002263.mm.1 TC1300000376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002262.mm.1 NM_001076790.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002262.mm.1 NM_001076790.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002262.mm.1 TC1300000376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300002264.mm.1 NM_001076790.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002264.mm.1 NM_001076790.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300002264.mm.1 TC1300000376.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 764190 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2960483 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2960483 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline