Search
Search

Eomes| Gene Symbol: Eomes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164789.1 | NP_001158261.1 | 4 - 5 | 1793 | 104 | |
| NM_010136.3 | NP_034266.2 | 4 - 5 | 1793 | 104 | ||
| GenBank mRNA | AB031037.1 | - | 4 - 5 | 1765 | 104 | |
| AK043327.1 | - | 4 - 5 | 1124 | 104 | ||
| AK089817.1 | - | 4 - 5 | 1353 | 104 | ||
| AK134951.1 | - | 1147 | 104 | |||
| AK143454.1 | - | 4 - 5 | 1114 | 104 | ||
| BC094319.1 | - | 4 - 5 | 1638 | 5721456 | 104 | |
Set Membership: |
|