Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01352089_m1
          See other LARP1 GE Assays ›
          Gene Symbol:
          Larp1
          Gene Name:
          La ribonucleoprotein domain family, member 1
          Gene Aliases:
          1810024J12Rik, 3110040D16Rik, Larp, mKIAA0731
          Chromosome Location:
          Chr.11: 58009064 - 58062034 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01352089_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Larp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028451.1 NP_082727.1 18 - 19 3137 71
          GenBank mRNA AK129202.1 - 7 - 8 885 71
          BC063078.1 - 5 - 6 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000674.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translational initiation
          cell proliferation
          positive regulation of macroautophagy
          TOR signaling
          positive regulation of translation
          RNA cap binding
          RNA binding
          mRNA 3'-UTR binding
          eukaryotic initiation factor 4E binding
          translation activator activity
          translation initiation factor binding
          poly(A) RNA binding
          mRNA 5'-UTR binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960456 AK169738.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960456 BC063078.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960456 NM_028451.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960456 AK007597.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P418908 BC053449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P418908 BC063078.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P418908 NM_028451.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P418908 AK129202.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P418908 BC053449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P418908 BC063078.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P418908 NM_028451.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P418908 AK129202.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 698669 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457347 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 437755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 171891 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 892992 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 112605 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380394 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876645 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 766131 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500939 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 534610 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 170987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 545385 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394178 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667288 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377878 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 401049 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 595810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 873172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 287371 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924192 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 938343 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 38167 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16634 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 744 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 46091 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165562 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960456 AK169738.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960456 BC063078.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960456 NM_028451.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960456 AK007597.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P418908 BC053449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P418908 BC063078.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P418908 NM_028451.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P418908 AK129202.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P418908 BC053449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P418908 BC063078.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P418908 NM_028451.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P418908 AK129202.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165562 - -
          Whole Mouse Genome G4122A A_52_P142170 AK169738.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P142170 BC063078.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P142170 NM_028451.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P142170 AK007597.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P12263 AK138840.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P12263 NM_028451.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418908 BC053449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418908 BC063078.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418908 NM_028451.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418908 AK129202.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418908 BC053449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418908 BC063078.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418908 NM_028451.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418908 AK129202.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P361112 BC063078.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P361112 NM_028451.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P361112 AK129202.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P772918 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27829017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09358613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09478811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09697422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09314808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09343498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27829003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27829021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09299871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09207764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08985902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09055184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09609955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09363486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09083800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08985903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09299852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09299829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27829001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09702902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09702897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09299843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09421320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09609961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09207757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09421325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09212773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09207758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09421331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27829018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09741370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27829022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09299851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09674184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09299866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09207777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09228529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27828984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09421326 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10376301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376299 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376308 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376305 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376302 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376303 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376310 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376295 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376309 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376298 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376294 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376311 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376296 - -
          Affymetrix HT MG-430 PM Array Plate 1428244_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10376312 - -
          Affymetrix Mouse Gene 1.1 ST Array 10376292 - -
          Mouse Genome 430 2.0 Array 1428244_PM_at - -
          Mouse Genome 430 2.0 Array 1428244_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73158_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73158_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100005398.mm.1 NM_028451.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005398.mm.1 NM_028451.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005398.mm.1 TC1100000674.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100005399.mm.1 NM_028451.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005399.mm.1 NM_028451.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005399.mm.1 TC1100000674.mm.1 MTA TC
          Mouse Expression Set 430 1428244_at - -
          Mouse Expression Set 430 1428244_PM_at - -
          Murine Genome U74v2 Set 94260_at - -
          Murine Genome U74v2 Set 94260_PM_at - -
          Mouse Genome 430A 2.0 Array 1428244_at - -
          Mouse Genome 430A 2.0 Array 1428244_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428244_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428244_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 902319 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline