Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01352377_m1
          See other LRRC14B GE Assays ›
          Gene Symbol:
          Lrrc14b
          Gene Name:
          leucine rich repeat containing 14B
          Gene Aliases:
          AI595366
          Chromosome Location:
          Chr.13: 74359582 - 74364000 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01352377_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc14b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033042.3 NP_001028214.1 1 - 2 935 68
          GenBank mRNA AK146537.1 - 1 - 2 935 68
          BC064819.1 - 1 - 2 940 68
          BC139119.1 - 1 - 2 914 8862141 68
          BC139120.1 - 1 - 2 914 8862142 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002319.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lrrc14b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052824 AK146537.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052824 BC064819.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052824 NM_001033042.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052824 BC049099.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052824 NM_001168659.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375723 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118277 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052824 AK146537.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052824 BC064819.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052824 NM_001033042.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052824 BC049099.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052824 NM_001168659.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29145868 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02722334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29145803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10636174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10966208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10966222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11121371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11121377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10966214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10966213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11121372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10785677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11121368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10785678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11121357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11121365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29145894 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10410642 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410643 - -
          Affymetrix Mouse Gene 1.1 ST Array 10410641 - -
          Affymetrix Mouse Clariom S Array PSR1300015274.mm.1 NM_001033042.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300015274.mm.1 NM_001033042.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300015274.mm.1 TC1300002319.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300015273.mm.1 NM_001033042.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300015273.mm.1 NM_001033042.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300015273.mm.1 TC1300002319.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300015272.mm.1 NM_001033042.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300015272.mm.1 NM_001033042.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300015272.mm.1 TC1300002319.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 852581 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161863 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161863 - -
          mouse-6 v1.1 (Illumina) GI_74315895-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161863 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161863 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline