Search
Search

Trip13| Gene Symbol: Trip13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027182.2 | NP_081458.1 | 5 - 6 | 723 | 114 | |
| GenBank mRNA | AK010336.1 | - | 5 - 6 | 685 | 114 | |
| AK142463.1 | - | 696 | 114 | |||
| AK146877.1 | - | 5 - 6 | 724 | 114 | ||
| AK151568.1 | - | 5 - 6 | 659 | 114 | ||
| BC064315.1 | - | 5 - 6 | 646 | 114 | ||
| BC126946.1 | - | 5 - 6 | 622 | 40090292 | 114 | |
Set Membership: |
|