Search
Search
| Gene Symbol: Drd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010076.3 | NP_034206.1 | 1 - 2 | 102 | 65 | |
| GenBank mRNA | AK044723.1 | - | 1 - 2 | 103 | 65 | |
| BY287823.1 | - | 1 - 2 | 150 | 65 | ||
Set Membership: |
|