Search
Search

Ifi30| Gene Symbol: Ifi30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023065.3 | NP_075552.2 | 3 - 4 | 417 | 68 | |
| GenBank mRNA | AF290973.1 | - | 3 - 4 | 418 | 68 | |
| AF309649.1 | - | 3 - 4 | 385 | 68 | ||
| AK156914.1 | - | 3 - 4 | 418 | 68 | ||
| BC054852.1 | - | 3 - 4 | 403 | 6528891 | 68 | |
| BE951500.1 | - | 4 - 5 | 68 | |||
| BQ746455.1 | - | 3 - 4 | 373 | 68 | ||
Set Membership: |
|