Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01545144_m1
          See other RFXAP GE Assays ›
          Gene Symbol:
          Rfxap
          Gene Name:
          regulatory factor X-associated protein
          Gene Aliases:
          5730495K23Rik
          Chromosome Location:
          Chr.3: 54803115 - 54807791 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01545144_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rfxap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133231.2 NP_573494.1 2 - 3 689 103
          GenBank mRNA AF335512.1 - 2 - 3 669 103
          AK077640.1 - 2 - 3 687 103
          AK087489.1 - 2 - 3 690 103
          AK144633.1 - 2 - 3 676 103
          AW826111.1 - 1 - 2 103
          BC019935.1 - 2 - 3 652 5041039 103
          BC047123.1 - 2 - 3 652 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001989.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rfxap

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of transcription, DNA-templated
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149939 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149942 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149943 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149940 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149944 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149937 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149936 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149941 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149938 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P441044 AK087489.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P441044 NM_133231.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P441044 AK087489.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P441044 NM_133231.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 83256 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82847 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136056 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P441044 AK087489.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P441044 NM_133231.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P441044 AK087489.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P441044 NM_133231.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124477 BC047123.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124477 BX519947.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124477 AK077640.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124477 AK144633.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124477 AW826111.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149949 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136056 - -
          Whole Mouse Genome G4122A A_51_P124477 BC047123.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124477 BX519947.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124477 AK077640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124477 AK144633.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124477 AW826111.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108737 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136056 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02744342 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02744328 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02548709 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02500885 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149943 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149939 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149944 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149937 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149941 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149940 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149936 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149942 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02845381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02567879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02744343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02744328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02744358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03118025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03136029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149906 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10498182 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498184 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498183 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498181 - -
          Affymetrix HT MG-430 PM Array Plate 1455303_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1418859_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10498180 - -
          Mouse Genome 430 2.0 Array 1455303_at - -
          Mouse Genome 430 2.0 Array 1418859_PM_at - -
          Mouse Genome 430 2.0 Array 1455303_PM_at - -
          Mouse Genome 430 2.0 Array 1418859_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170767_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170767_at - -
          Affymetrix Mouse Clariom S Array PSR0300015150.mm.1 NM_133231.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300015150.mm.1 NM_133231.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300015150.mm.1 TC0300001989.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300015151.mm.1 NM_133231.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300015151.mm.1 NM_133231.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300015151.mm.1 TC0300001989.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300015152.mm.1 NM_133231.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300015152.mm.1 NM_133231.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300015152.mm.1 TC0300001989.mm.1 MTA TC
          Mouse Expression Set 430 1418859_PM_at - -
          Mouse Expression Set 430 1418859_at - -
          Mouse Expression Set 430 1455303_at - -
          Mouse Expression Set 430 1455303_PM_at - -
          Mouse Genome 430A 2.0 Array 1418859_PM_at - -
          Mouse Genome 430A 2.0 Array 1418859_at - -
          Affymetrix HT Mouse Genome 430A Array 1418859_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418859_at - -
          Affymetrix HT Mouse Genome 430B Array 1455303_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455303_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106020524 - -
          MouseWG-6 v2 Expression BeadChip 3290181 - -
          Illumina mouse-6 v1.0 expression beadchip 106020524 - -
          Illumina mouse-6 v1.0 expression beadchip 3290181 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003290181 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2644992 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2443602 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2644992 - -
          mouse-6 v1.1 (Illumina) scl00170767.2_151-S - -
          mouse-6 v1.1 (Illumina) scl22157.4_24-S - -
          Illumina Sentrix mouse-6.v1 bead array 106020524 - -
          Illumina Sentrix mouse-6.v1 bead array 3290181 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00170767.2_151-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3290181 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106020524 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2644992 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2644992 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00170767.2_151-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline