Search
Search

Skor1| Gene Symbol: Skor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163755.1 | NP_001157227.1 | 6 - 7 | 2665 | 69 | |
| NM_001163757.1 | NP_001157229.1 | 6 - 7 | 2665 | 69 | ||
| NM_001163758.1 | NP_001157230.1 | 6 - 7 | 2646 | 69 | ||
| NM_172446.3 | NP_766034.2 | 6 - 7 | 2718 | 69 | ||
| GenBank mRNA | AB185113.1 | - | 6 - 7 | 2668 | 69 | |
| AK029916.1 | - | 6 - 7 | 2649 | 69 | ||
| AK049035.1 | - | 6 - 7 | 2719 | 69 | ||
| AK134840.1 | - | 6 - 7 | 2657 | 69 | ||
| BC138063.1 | - | 6 - 7 | 2622 | 8861082 | 69 | |
Set Membership: |
|