Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01599204_g1
          See other 1500009L16RIK GE Assays ›
          Gene Symbol:
          1500009L16Rik
          Gene Name:
          RIKEN cDNA 1500009L16 gene
          Gene Aliases:
          2700050C19Rik, OCC-1
          Chromosome Location:
          Chr.10: 83722666 - 83762762 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01599204_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1500009L16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145198.1 NP_001138670.1 5 - 6 355 70
          GenBank mRNA AK005183.1 - 5 - 6 286 70
          BC048550.1 - 5 - 6 355 70

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126790 AK012407.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126790 CD776925.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126790 CO044092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126790 NM_001145198.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 260790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 627814 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812736 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906693 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 21808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 780834 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 712337 NM_001011757.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262523 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385497 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597447 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 796777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593941 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 57356 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297280 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511882 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535651 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 671788 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584264 AF082348.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584264 AJ010259.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584264 AJ132406.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584264 AK028735.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584264 AK135757.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584264 AK136127.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584264 AK136129.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584264 AK139773.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584264 AK139847.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584264 AK139936.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584264 BC055363.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584264 NM_009757.4 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916592 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 608125 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 6220 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133928 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 50132 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 162797 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126790 AK012407.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126790 CD776925.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126790 CO044092.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126790 NM_001145198.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02173635 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02173653 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07129816 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102236 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02173653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08399720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08484204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08901364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08849954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08335750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08338743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08565729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08338738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08338740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08942410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08484200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27298945 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1442649_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1452840_PM_at - -
          Mouse Genome 430 2.0 Array 1442649_at - -
          Mouse Genome 430 2.0 Array 1452840_at - -
          Mouse Genome 430 2.0 Array 1452840_PM_at - -
          Mouse Genome 430 2.0 Array 1442649_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69784_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69784_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000007624.mm.1 NM_001145198.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007624.mm.1 NM_001145198.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007624.mm.1 TC1000001011.mm.1 MTA TC
          Mouse Expression Set 430 1442649_PM_at - -
          Mouse Expression Set 430 1442649_at - -
          Mouse Expression Set 430 1452840_PM_at - -
          Mouse Expression Set 430 1452840_at - -
          Affymetrix HT Mouse Genome 430B Array 1452840_at - -
          Affymetrix HT Mouse Genome 430B Array 1442649_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442649_at - -
          Affymetrix HT Mouse Genome 430B Array 1452840_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 355364 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106200736 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248734 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline