Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01603319_g1
          See other LIPO1 GE Assays ›
          Gene Symbol:
          Lipo1
          Gene Name:
          lipase, member O1
          Gene Aliases:
          AI747699
          Chromosome Location:
          Chr.19: 33555160 - 33591172 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01603319_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lipo1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013770.3 NP_001013792.2 10 - 11 1526 84
          GenBank mRNA AK135585.1 - 4 - 5 677 84
          AK139790.1 - 10 - 11 1527 84
          AK170332.1 - 10 - 11 1460 84
          BC052506.1 - 3 - 4 281 84
          BC145463.1 - 3 - 4 242 84
          EU234002.1 - 3 - 4 2106 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          hydrolase lipase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040777 BC052506.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040777 AK139790.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040777 AK170332.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040777 NM_001013770.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354519 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45925 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040777 BC052506.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040777 AK139790.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040777 AK170332.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040777 NM_001013770.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09156442 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03586523 - -
          Whole Mouse Genome G4122A A_52_P476454 AK139790.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P476454 AK170332.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P476454 NM_001013770.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58594 BC052506.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58594 AK170332.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09156400 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03586527 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14168299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32076257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32076253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32076264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32076266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32076254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32076256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32076259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32076367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14475342 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10467114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467112 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467113 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467111 - -
          Affymetrix HT MG-430 PM Array Plate 1433914_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10467110 - -
          Mouse Genome 430 2.0 Array 1433914_PM_at - -
          Mouse Genome 430 2.0 Array 1431151_at - -
          Mouse Genome 430 2.0 Array 1433914_at - -
          Mouse Genome 430 2.0 Array 1431151_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381236_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381236_PM_at - -
          Mouse Expression Set 430 1431151_at - -
          Mouse Expression Set 430 1431151_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433914_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433914_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 482117 - -
          ABI Mouse Genome Survey Microarray 710407 - -
          ABI Mouse Genome Survey Microarray 835299 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162814 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162814 - -
          mouse-6 v1.1 (Illumina) GI_85701463-I - -
          MouseRef-8 v2 Expression BeadChip ILMN_2793739 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2793739 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) GI_85701463-I - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline