Search
Search

Hdac1| Gene Symbol: Hdac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008228.2 | NP_032254.1 | 7 - 8 | 749 | 78 | |
| GenBank mRNA | AK076080.1 | - | 7 - 8 | 734 | 78 | |
| BC092070.1 | - | 7 - 8 | 744 | 4217199 | 78 | |
| BC092285.1 | - | 7 - 8 | 749 | 78 | ||
| BC108371.1 | - | 7 - 8 | 750 | 4976514 | 78 | |
| U80780.1 | - | 7 - 8 | 746 | 78 | ||
| X98207.1 | - | 7 - 8 | 749 | 78 | ||
Set Membership: |
|