| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600016745.mm.1 |
NM_018810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016745.mm.1 |
NM_018810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016745.mm.1 |
TC0600002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016760.mm.1 |
NM_018810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016760.mm.1 |
NM_018810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016760.mm.1 |
TC0600002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016746.mm.1 |
NM_018810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016746.mm.1 |
NM_018810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016746.mm.1 |
TC0600002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016754.mm.1 |
NM_018810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016754.mm.1 |
NM_018810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016754.mm.1 |
TC0600002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016767.mm.1 |
NM_018810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016767.mm.1 |
NM_018810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016767.mm.1 |
TC0600002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016753.mm.1 |
NM_018810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016753.mm.1 |
NM_018810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016753.mm.1 |
TC0600002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016755.mm.1 |
NM_018810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016755.mm.1 |
NM_018810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016755.mm.1 |
TC0600002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016759.mm.1 |
NM_018810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016759.mm.1 |
NM_018810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016759.mm.1 |
TC0600002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016743.mm.1 |
NM_018810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016743.mm.1 |
NM_018810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016743.mm.1 |
TC0600002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016750.mm.1 |
TC0600002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016751.mm.1 |
NM_018810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016751.mm.1 |
NM_018810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016751.mm.1 |
TC0600002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016752.mm.1 |
NM_018810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016752.mm.1 |
NM_018810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016752.mm.1 |
TC0600002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016766.mm.1 |
NM_018810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016766.mm.1 |
NM_018810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016766.mm.1 |
TC0600002122.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016744.mm.1 |
NM_018810.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016744.mm.1 |
NM_018810.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016744.mm.1 |
TC0600002122.mm.1 |
MTA TC |