Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01612741_m1
          See other LYZ2 GE Assays ›
          Gene Symbol:
          Lyz2
          Gene Name:
          lysozyme 2
          Gene Aliases:
          AI326280, Lys, Lysm, Lyzs, Lzm, Lzm-s1, Lzp
          Chromosome Location:
          Chr.10: 117277541 - 117282272 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01612741_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lyz2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017372.3 NP_059068.1 1 - 2 166 134
          GenBank mRNA AK148516.1 - 1 - 2 167 134
          AK150998.1 - 1 - 2 169 134
          AK153244.1 - 1 - 2 169 134
          AK153475.1 - 1 - 2 167 134
          AK159276.1 - 1 - 2 216 134
          AK159640.1 - 1 - 2 163 134
          BC002069.1 - 1 - 2 158 3593360 134
          BC054463.1 - 1 - 2 158 6514977 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002929.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lyz2

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          cytolysis
          defense response to bacterium
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          lysozyme activity
          catalytic activity
          hydrolase activity
          hydrolase activity, acting on glycosyl bonds
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P321150 BC002069.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P321150 AK159640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P321150 BC054463.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P321150 AK150998.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P321150 AK153244.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P321150 BC002069.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P321150 AK159640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P321150 BC054463.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P321150 AK150998.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P321150 AK153244.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915695 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971572 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 80656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768688 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 105589 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37343 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P321150 BC002069.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P321150 AK159640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P321150 BC054463.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P321150 AK150998.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P321150 AK153244.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P321150 BC002069.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P321150 AK159640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P321150 BC054463.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P321150 AK150998.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P321150 AK153244.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P321150 BC002069.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P321150 AK159640.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P321150 BC054463.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P321150 AK150998.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P321150 AK153244.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P321150 BC002069.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P321150 AK159640.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P321150 BC054463.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P321150 AK150998.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P321150 AK153244.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P238027 BC002069.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P238027 BC019611.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P238027 BC054463.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P238027 AK150998.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P238027 AK153244.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27480346 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07206589 - -
          Whole Mouse Genome G4122A A_52_P238027 BC002069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P238027 BC019611.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P238027 BC054463.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P238027 AK150998.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P238027 AK153244.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P321150 BC002069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P321150 AK159640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P321150 BC054463.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P321150 AK150998.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P321150 AK153244.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P321150 BC002069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P321150 AK159640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P321150 BC054463.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P321150 AK150998.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P321150 AK153244.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127013 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126294 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109188 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08373585 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08373596 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08373598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27480396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08597900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08308267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27480366 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10372650 - -
          Affymetrix Mouse Gene 1.0 ST Array 10372649 - -
          Affymetrix Mouse Gene 1.0 ST Array 10372651 - -
          Affymetrix HT MG-430 PM Array Plate 1423547_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10372648 - -
          Mouse Genome 430 2.0 Array 1423547_at - -
          Mouse Genome 430 2.0 Array 1423547_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17105_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17105_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000022139.mm.1 NM_017372.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022139.mm.1 NM_017372.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022139.mm.1 TC1000002929.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000022137.mm.1 NM_017372.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022137.mm.1 NM_017372.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022137.mm.1 TC1000002929.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000022136.mm.1 NM_017372.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022136.mm.1 NM_017372.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022136.mm.1 TC1000002929.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000022138.mm.1 NM_017372.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022138.mm.1 NM_017372.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000022138.mm.1 TC1000002929.mm.1 MTA TC
          Mouse Expression Set 430 1450822_PM_at - -
          Mouse Expression Set 430 1450822_at - -
          Mouse Expression Set 430 1423547_at - -
          Mouse Expression Set 430 1423547_PM_at - -
          Murine Genome U74v2 Set 100611_at - -
          Murine Genome U74v2 Set 162309_PM_at - -
          Murine Genome U74v2 Set 101753_PM_s_at - -
          Murine Genome U74v2 Set 100611_PM_at - -
          Murine Genome U74v2 Set 162309_at - -
          Murine Genome U74v2 Set 101753_s_at - -
          Mouse Genome 430A 2.0 Array 1423547_PM_at - -
          Mouse Genome 430A 2.0 Array 1423547_at - -
          Mouse Genome 430A 2.0 Array 1450822_PM_at - -
          Mouse Genome 430A 2.0 Array 1450822_at - -
          Affymetrix HT Mouse Genome 430A Array 1423547_at - -
          Affymetrix HT Mouse Genome 430A Array 1423547_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 374617 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105340427 - -
          Illumina mouse-6 v1.0 expression beadchip 105340427 - -
          Illumina mouse-6 v1.0 expression beadchip 6770717 - -
          Illumina mouse-6 v1.0 expression beadchip 50068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000050068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770717 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1252076 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2766780 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252076 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2766780 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2453243 - -
          mouse-6 v1.1 (Illumina) scl5225.1.1_39-S - -
          mouse-6 v1.1 (Illumina) scl0017105.1_34-S - -
          mouse-6 v1.1 (Illumina) scl0017105.2_305-S - -
          Illumina Sentrix mouse-6.v1 bead array 50068 - -
          Illumina Sentrix mouse-6.v1 bead array 105340427 - -
          Illumina Sentrix mouse-6.v1 bead array 6770717 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0017105.1_34-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0017105.2_305-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 50068 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105340427 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770717 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2766780 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1252076 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2939681 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2939681 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252076 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2766780 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0017105.1_34-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline