Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01617780_m1
          See other VMN2R12 GE Assays ›
          Gene Symbol:
          Vmn2r12
          Gene Name:
          vomeronasal 2, receptor 12
          Gene Aliases:
          EG627569, Gm6769
          Chromosome Location:
          Chr.5: 109085849 - 109097864 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01617780_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn2r12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001104623.1 NP_001098093.1 3 - 4 1293 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002963.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035394 NM_001104623.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035394 NM_001104623.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23975066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23975055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23975125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23975087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23975160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04108968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04340288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23975125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23975123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04844863 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10532279 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532278 - -
          Affymetrix Mouse Gene 1.1 ST Array 10532277 - -
          Affymetrix Mouse Clariom S Array PSR0500026229.mm.1 NM_001104623.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026229.mm.1 NM_001104623.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026229.mm.1 TC0500002963.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026230.mm.1 NM_001104623.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026230.mm.1 NM_001104623.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026230.mm.1 TC0500002963.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026228.mm.1 NM_001104623.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026228.mm.1 NM_001104623.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026228.mm.1 TC0500002963.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026231.mm.1 NM_001104623.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026231.mm.1 NM_001104623.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026231.mm.1 TC0500002963.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026226.mm.1 NM_001104623.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026226.mm.1 NM_001104623.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026226.mm.1 TC0500002963.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026227.mm.1 NM_001104623.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026227.mm.1 NM_001104623.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026227.mm.1 TC0500002963.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 876510 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline