Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01618651_s1
          See other OLFR111 GE Assays ›
          Gene Symbol:
          Olfr111
          Gene Name:
          olfactory receptor 111
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 37504960 - 37536128 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01618651_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr111
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005485.2 NP_001005485.1 1 483 67
          GenBank mRNA BC139116.1 - 1 483 8862138 67
          BC139117.1 - 1 483 8862139 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr111

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163368 NM_001005485.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163368 BC139116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163368 BC139117.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163368 NM_001005485.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163368 BC139116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163368 BC139117.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P345011 NM_001005485.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P345011 BC139116.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P345011 BC139117.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P345011 BC132347.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P345011 NM_146328.2 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13367730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13367737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13367744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13343804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13311233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13367746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13205058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169074 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445142 - -
          Affymetrix Mouse Gene 1.1 ST Array 10445141 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2967322 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2967322 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline