Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01621586_s1
          See other CTLA2B GE Assays ›
          Gene Symbol:
          Ctla2b
          Gene Name:
          cytotoxic T lymphocyte-associated protein 2 beta
          Gene Aliases:
          AW545568, Ctla-2b
          Chromosome Location:
          Chr.13: 60895351 - 60897932 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01621586_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctla2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145801.1 NP_001139273.1 4 1002 107
          NM_007797.1 NP_031823.1 5 701 107
          GenBank mRNA AK150917.1 - 4 961 107
          AY034578.1 - 5 641 107
          DT925153.1 - 463 107
          X15592.1 - 5 641 107

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endopeptidase activity
          proteolysis involved in cellular protein catabolic process
          cysteine-type endopeptidase activity
          cysteine-type endopeptidase inhibitor activity
          protein binding
          cysteine-type peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P489522 AK150917.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P489522 AY034578.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P489522 BE448128.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P489522 NM_007797.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P489522 DT929456.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P489522 AK150917.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P489522 AY034578.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P489522 BE448128.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P489522 NM_007797.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P489522 DT929456.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137766 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P489522 AK150917.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P489522 AY034578.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P489522 BE448128.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P489522 NM_007797.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P489522 DT929456.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P489522 AK150917.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P489522 AY034578.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P489522 BE448128.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P489522 NM_007797.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P489522 DT929456.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137766 - -
          Whole Mouse Genome G4122A A_51_P489522 AK150917.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489522 AY034578.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489522 BE448128.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489522 NM_007797.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489522 DT929456.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489522 AK150917.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489522 AY034578.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489522 BE448128.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489522 NM_007797.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P489522 DT929456.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087925 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10409868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409867 - -
          Affymetrix HT MG-430 PM Array Plate 1452352_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10409866 - -
          Mouse Genome 430 2.0 Array 1452352_at - -
          Mouse Genome 430 2.0 Array 1452352_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300013738.mm.1 NM_001145801.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013738.mm.1 NM_001145801.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013738.mm.1 NM_007797.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013738.mm.1 NM_007797.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013738.mm.1 TC1300002141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013739.mm.1 NM_001145801.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013739.mm.1 NM_001145801.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013739.mm.1 TC1300002141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013742.mm.1 NM_001145801.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013742.mm.1 NM_001145801.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013742.mm.1 NM_007797.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013742.mm.1 NM_007797.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013742.mm.1 TC1300002141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013743.mm.1 NM_001145801.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013743.mm.1 NM_001145801.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013743.mm.1 NM_007797.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013743.mm.1 NM_007797.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013743.mm.1 TC1300002141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013741.mm.1 NM_001145801.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013741.mm.1 NM_001145801.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013741.mm.1 NM_007797.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013741.mm.1 NM_007797.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013741.mm.1 TC1300002141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013740.mm.1 NM_001145801.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013740.mm.1 NM_001145801.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013740.mm.1 NM_007797.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013740.mm.1 NM_007797.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013740.mm.1 TC1300002141.mm.1 MTA TC
          Mouse Expression Set 430 1452352_at - -
          Mouse Expression Set 430 1452352_PM_at - -
          Mouse Expression Set 430 1416811_PM_s_at - -
          Mouse Expression Set 430 1416811_s_at - -
          Murine Genome U74v2 Set 103518_at - -
          Murine Genome U74v2 Set 96912_PM_s_at - -
          Murine Genome U74v2 Set 103518_PM_at - -
          Murine Genome U74v2 Set 96912_s_at - -
          Mouse Genome 430A 2.0 Array 1452352_at - -
          Mouse Genome 430A 2.0 Array 1452352_PM_at - -
          Mouse Genome 430A 2.0 Array 1416811_s_at - -
          Mouse Genome 430A 2.0 Array 1416811_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452352_at - -
          Affymetrix HT Mouse Genome 430A Array 1452352_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 355535 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline