Search
Search

Usp17la| Gene Symbol: Usp17la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007887.2 | NP_031913.1 | 3 | 870 | 135 | |
| GenBank mRNA | AY148453.1 | - | 2 | 699 | 135 | |
| U41636.1 | - | 3 | 882 | 135 | ||
Set Membership: |
|