Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01701109_m1
          See other APOL9B GE Assays ›
          Gene Symbol:
          Apol9b Apol9a
          Gene Name:
          apolipoprotein L 9b
          apolipoprotein L 9a
          Gene Aliases:
          2310016F22Rik
          -
          Chromosome Location:
          Chr.15: 77729072 - 77736382 on Build GRCm38
          Chromosome Location:
          Chr.15: 77403789 - 77411080 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm01701109_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apol9b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173743.4 NP_776104.1 1 - 2 73 55
          GenBank mRNA AK050167.1 - 1 - 2 75 55
          Gene Symbol: Apol9a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001162883.1 NP_001156355.1 1 - 2 51 55
          GenBank mRNA CJ058381.1 - 1 - 2 52 55

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK050167.1, NM_001162883.1, NM_173743.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apol9b
          Click here to find siRNAs for Gene Apol9a

          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110633 AK050167.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110633 BF148017.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110633 NM_001168660.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110633 NM_173743.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043367 NM_173786.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043367 NM_001162883.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043367 BC020489.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107775 BC057684.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107775 NM_173786.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107775 NM_001162883.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107775 BC020489.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107775 AK050167.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295916 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170727 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168088 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110633 AK050167.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110633 BF148017.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110633 NM_001168660.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110633 NM_173743.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107775 BC057684.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107775 NM_173786.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107775 NM_001162883.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107775 BC020489.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107775 AK050167.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043367 NM_173786.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043367 NM_001162883.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043367 BC020489.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30355337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30355387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30355398 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03075616 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168088 - -
          Whole Mouse Genome G4122A A_52_P803082 - -
          Whole Mouse Genome G4122A A_51_P500215 AK050167.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500215 BF148017.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500215 NM_001168660.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500215 NM_173743.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500215 AK009386.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131188 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110633 AK050167.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110633 BF148017.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110633 NM_001168660.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110633 NM_173743.4 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168088 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12114329 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12114343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30355476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30355398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30355477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30355400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30355410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30355474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30355471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30355446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30355473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12177540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12198344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30354008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12198347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30355370 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10430176 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425050 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430175 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430178 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425050 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430176 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430175 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430178 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430177 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425051 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425052 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425052 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425051 - -
          Affymetrix Mouse Gene 1.1 ST Array 10425049 - -
          Affymetrix Mouse Gene 1.1 ST Array 10430174 - -
          Affymetrix Mouse Gene 1.1 ST Array 10430174 - -
          Affymetrix Mouse Gene 1.1 ST Array 10425049 - -
          Mouse Genome 430 2.0 Array 1424518_PM_at - -
          Mouse Genome 430 2.0 Array 1424518_PM_at - -
          Mouse Genome 430 2.0 Array 1424518_at - -
          Mouse Genome 430 2.0 Array 1424518_at - -
          Affymetrix Mouse Clariom S Array PSR1500014048.mm.1 NM_001162883.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014048.mm.1 NM_001162883.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014048.mm.1 NM_173786.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014048.mm.1 NM_173786.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014048.mm.1 TC1500001809.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004609.mm.1 NM_001168660.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004609.mm.1 NM_001168660.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004609.mm.1 NM_173743.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004609.mm.1 NM_173743.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004609.mm.1 TC1500000632.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014054.mm.1 NM_001162883.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014054.mm.1 NM_001162883.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014054.mm.1 TC1500001809.mm.1 MTA TC
          Mouse Expression Set 430 1424518_PM_at - -
          Mouse Expression Set 430 1424518_at - -
          Mouse Expression Set 430 1424518_at - -
          Mouse Expression Set 430 1424518_PM_at - -
          Mouse Genome 430A 2.0 Array 1424518_PM_at - -
          Mouse Genome 430A 2.0 Array 1424518_at - -
          Mouse Genome 430A 2.0 Array 1424518_PM_at - -
          Mouse Genome 430A 2.0 Array 1424518_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 358743 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2350438 - -
          Illumina mouse-6 v1.0 expression beadchip 2350438 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002350438 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2671434 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2622856 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2622856 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2671434 - -
          mouse-6 v1.1 (Illumina) scl0071898.1_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 2350438 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0071898.1_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2350438 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2622856 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2622856 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline