Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01701878_s1
          See other GM5136 GE Assays ›
          Gene Symbol:
          Gm5136
          Gene Name:
          predicted gene 5136
          Gene Aliases:
          C6.1AL, EG368203
          Chromosome Location:
          Chr.10: 108699044 - 108700160 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01701878_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm5136
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_203660.2 NP_988991.1 1 561 104
          GenBank mRNA AK134982.1 - 1 91 104
          BC120506.1 - 1 561 40129429 104
          BC120508.1 - 1 561 40129431 104

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gm5136

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          biological_process
          molecular_function
          peptidase activity
          metallopeptidase activity
          hydrolase activity
          pyroglutamyl-peptidase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067645 NM_203660.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067645 AK134982.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067645 BC120506.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067645 BC120508.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137771 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067645 NM_203660.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067645 AK134982.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067645 BC120506.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067645 BC120508.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27433503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08777484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08664995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08273355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08846991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08777488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08273354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27433516 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10372339 - -
          Affymetrix Mouse Gene 1.1 ST Array 10372338 - -
          Affymetrix Mouse Clariom S Array PSR1000021554.mm.1 NM_203660.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021554.mm.1 NM_203660.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021554.mm.1 TC1000002834.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000021555.mm.1 NM_203660.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021555.mm.1 NM_203660.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000021555.mm.1 TC1000002834.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1770324 - -
          Illumina mouse-6 v1.0 expression beadchip 1770324 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770324 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2669886 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2669886 - -
          mouse-6 v1.1 (Illumina) scl37528.1.1_99-S - -
          Illumina Sentrix mouse-6.v1 bead array 1770324 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37528.1.1_99-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770324 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2669886 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2669886 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline