Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01702043_s1
          See other TAS2R118 GE Assays ›
          Gene Symbol:
          Tas2r118
          Gene Name:
          taste receptor, type 2, member 118
          Gene Aliases:
          Gm465, T2R18, Tas2r16, Tas2r18, mGR18, mt2r40
          Chromosome Location:
          Chr.6: 23969161 - 23970060 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01702043_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tas2r118
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207022.1 NP_996905.1 1 849 89
          GenBank mRNA BC104400.2 - 1 841 89
          BC104401.1 - 1 849 40050472 89

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tas2r118

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          signal transduction
          G-protein coupled receptor signaling pathway
          response to stimulus
          sensory perception of taste
          signal transducer activity
          G-protein coupled receptor activity
          bitter taste receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P333825 NM_207022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P333825 BC104400.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P333825 BC104401.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P333825 NM_207022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P333825 BC104400.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P333825 BC104401.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129771 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P333825 NM_207022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P333825 BC104400.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P333825 BC104401.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P333825 NM_207022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P333825 BC104400.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P333825 BC104401.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P333825 NM_207022.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P333825 BC104400.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P333825 BC104401.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P333825 NM_207022.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P333825 BC104400.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P333825 BC104401.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24299375 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129771 - -
          Whole Mouse Genome G4122A A_52_P333825 NM_207022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P333825 BC104400.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P333825 BC104401.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P333825 NM_207022.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P333825 BC104400.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P333825 BC104401.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113696 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129771 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05326205 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05326189 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05326187 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05326191 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05326192 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05326210 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05326185 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05326193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24299368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05326185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05668295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05710672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05710667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05710704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05326204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05830337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05592387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05326192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05326199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24299375 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10543410 - -
          Affymetrix Mouse Gene 1.1 ST Array 10543409 - -
          Affymetrix Mouse Clariom S Array PSR0600015187.mm.1 NM_207022.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015187.mm.1 NM_207022.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015187.mm.1 TC0600001938.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3710025 - -
          Illumina mouse-6 v1.0 expression beadchip 3710025 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003710025 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1254787 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254787 - -
          mouse-6 v1.1 (Illumina) scl0387347.1_262-S - -
          Illumina Sentrix mouse-6.v1 bead array 3710025 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0387347.1_262-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3710025 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2706111 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2706111 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline