| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300010983.mm.1 |
NM_173051.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300010983.mm.1 |
NM_173051.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300010983.mm.1 |
TC1300001764.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300010980.mm.1 |
NM_173051.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300010980.mm.1 |
NM_173051.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300010980.mm.1 |
TC1300001764.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300010979.mm.1 |
NM_173051.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300010979.mm.1 |
NM_173051.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300010979.mm.1 |
TC1300001764.mm.1 |
MTA TC |