Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01703434_s1
          See other IFNA16 GE Assays ›
          Gene Symbol:
          Ifna16
          Gene Name:
          interferon alpha 16
          Gene Aliases:
          BC114392, Gm13280, Ifna6T, Ifnat6, OTTMUSG00000011275
          Chromosome Location:
          Chr.4: 88676287 - 88676856 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01703434_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifna16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_206867.1 NP_996750.1 1 236 97
          GenBank mRNA BC114392.1 - 1 235 97

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifna16

          Panther Classification:

          Molecular Function -

          interferon superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          natural killer cell activation involved in immune response
          humoral immune response
          cytokine-mediated signaling pathway
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          B cell proliferation
          response to exogenous dsRNA
          cytokine activity
          type I interferon receptor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049196 NM_206867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049196 BC114392.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049196 NM_206867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049196 BC114392.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091625 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03559822 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03840868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03771055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03717594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03840864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03682136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03840865 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10505893 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514314 - -
          Affymetrix Mouse Gene 1.1 ST Array 10514313 - -
          Affymetrix Mouse Clariom S Array PSR0400024332.mm.1 NM_206867.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024332.mm.1 NM_206867.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024332.mm.1 TC0400003011.mm.1 MTA TC
          Mouse Expression Set 430 1422403_PM_at - -
          Mouse Expression Set 430 1422403_at - -
          Mouse Expression Set 430 1422404_PM_x_at - -
          Mouse Expression Set 430 1422404_x_at - -
          Murine Genome U74v2 Set 102149_PM_f_at - -
          Murine Genome U74v2 Set 102149_f_at - -
          Mouse Genome 430A 2.0 Array 1422404_x_at - -
          Mouse Genome 430A 2.0 Array 1422403_PM_at - -
          Mouse Genome 430A 2.0 Array 1422403_at - -
          Mouse Genome 430A 2.0 Array 1422404_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6980044 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980044 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2742656 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2742656 - -
          mouse-6 v1.1 (Illumina) scl0230398.1_10-S - -
          Illumina Sentrix mouse-6.v1 bead array 6980044 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0230398.1_10-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980044 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2742656 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2742656 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline