Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01705338_s1
          See other ISG15 GE Assays ›
          Gene Symbol:
          Isg15
          Gene Name:
          ISG15 ubiquitin-like modifier
          Gene Aliases:
          G1p2, IGI15, IP17, Irfp, UCRP
          Chromosome Location:
          Chr.4: 156199424 - 156200818 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01705338_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Isg15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
          RefSeq NM_015783.3 NP_056598.2 2 346 107
          GenBank mRNA BC031424.1 - 2 313 107
          BC083156.1 - 2 295 107
          BC109346.2 - 2 296 107
          CT010333.1 - 1 236 107
          X56602.1 - 2 347 107

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Isg15

          Gene Ontology Categories:

          Function(s) Process(es)

          modification-dependent protein catabolic process
          negative regulation of protein ubiquitination
          ISG15-protein conjugation
          regulation of interferon-gamma production
          response to type I interferon
          defense response to bacterium
          negative regulation of viral genome replication
          positive regulation of erythrocyte differentiation
          defense response to virus
          protein binding
          protein tag

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103698 BC109346.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103698 X56602.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103698 NM_015783.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103698 BC031424.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2103698 BC083156.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128537 X56602.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128537 BC109346.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128537 CT010333.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128537 NM_015783.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128537 BC031424.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103698 BC109346.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103698 X56602.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103698 NM_015783.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103698 BC031424.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2103698 BC083156.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128537 X56602.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128537 BC109346.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128537 CT010333.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128537 NM_015783.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128537 BC031424.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31206673 - -
          Whole Mouse Genome G4122A A_52_P463936 BC109346.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P463936 CT010333.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P463936 X56602.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P463936 NM_015783.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P463936 BC031424.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128537 X56602.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128537 BC109346.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128537 CT010333.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128537 NM_015783.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128537 BC031424.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10451288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451290 - -
          Affymetrix Mouse Gene 1.1 ST Array 10451287 - -
          Mouse Genome 430 2.0 Array 1431591_s_at - -
          Mouse Genome 430 2.0 Array 1431591_PM_s_at - -
          Affymetrix Mouse Clariom S Array PSR0400035093.mm.1 NM_015783.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035093.mm.1 NM_015783.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035093.mm.1 TC0400004171.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400035098.mm.1 NM_015783.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035098.mm.1 NM_015783.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035098.mm.1 TC0400004171.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400035095.mm.1 NM_015783.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035095.mm.1 NM_015783.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035095.mm.1 TC0400004171.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400035094.mm.1 NM_015783.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035094.mm.1 NM_015783.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035094.mm.1 TC0400004171.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400035092.mm.1 NM_015783.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035092.mm.1 NM_015783.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035092.mm.1 TC0400004171.mm.1 MTA TC
          Mouse Expression Set 430 1431591_s_at - -
          Mouse Expression Set 430 1431591_PM_s_at - -
          Murine Genome U74v2 Set 98822_PM_at - -
          Murine Genome U74v2 Set 161511_PM_f_at - -
          Murine Genome U74v2 Set 161511_f_at - -
          Murine Genome U74v2 Set 98822_at - -
          Mouse Genome 430A 2.0 Array 1431591_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1431591_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 898210 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3170093 - -
          Illumina mouse-6 v1.0 expression beadchip 3170093 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003170093 - -
          mouse-6 v1.1 (Illumina) scl053606.2_17-S - -
          Illumina Sentrix mouse-6.v1 bead array 3170093 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl053606.2_17-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3170093 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256257 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256257 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl053606.2_17-S - -

          Back To Top
           

          Citations (1)

          Suppression of interferon (IFN)-inducible genes and IFN-mediated functional responses in BCR-ABL-expressing cells.
          Authors
          Katsoulidis E, Sassano A, Majchrzak-Kita B, Carayol N, Yoon P, Jordan A, Druker BJ, Fish EN, Platanias LC
          Journal
          J Biol Chem - Page No 10793 (Fri Apr 18 00:00:00 UTC 2008)
          Volume (Year)
          283 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch