Search
Search

Klhl7| Gene Symbol: Klhl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035420.1 | - | 1 | 1354 | 122 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 126, member A |
Fam126a |
AB030242, Drctnnb1a |
-130.6kb | Anti-sense | Chr. 5 - 23915276 - 24030780 on NCBI Build GRCm38 |
| Downstream | nucleoporin like 2 | Nupl2 |
AV116043, CG1, NLP-1, Nlp1 |
1.3kb | Sense | Chr. 5 - 24164700 - 24184008 on NCBI Build GRCm38 |
Set Membership: |
|