Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01745688_s1
          See other SKA2 GE Assays ›
          Gene Symbol:
          Ska2
          Gene Name:
          spindle and kinetochore associated complex subunit 2
          Gene Aliases:
          1110001A07Rik, C78640, Fam33a
          Chromosome Location:
          Chr.11: 87109261 - 87122974 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01745688_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ska2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025377.3 NP_079653.1 4 470 155
          GenBank mRNA AK003196.1 - 4 498 155
          AK013317.1 - 4 471 155
          AK013370.1 - 4 471 155
          AK017811.1 - 4 471 155
          AK162628.1 - 467 155
          BC052151.1 - 471 155
          BC060088.1 - 473 155
          BC096627.1 - 482 155
          BC117001.1 - 4 500 40126320 155
          BC119167.1 - 4 500 8733916 155

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ska2

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic anaphase
          cell cycle
          chromosome segregation
          mitotic nuclear division
          regulation of microtubule polymerization or depolymerization
          cell division
          microtubule binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27992598 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27992601 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27992600 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P237512 BC119167.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P237512 BC060088.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P237512 BC096627.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P237512 BC117001.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P237512 AK162628.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P237512 BC119167.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P237512 BC060088.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P237512 BC096627.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P237512 BC117001.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P237512 AK162628.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 134039 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390526 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 411159 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 761123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 91341 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 109880 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 875172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600097 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 689534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824201 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42790 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110209 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P237512 BC119167.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P237512 BC060088.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P237512 BC096627.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P237512 BC117001.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P237512 AK162628.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P237512 BC119167.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P237512 BC060088.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P237512 BC096627.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P237512 BC117001.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P237512 AK162628.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27992669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27992631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27992668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27992630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27992676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27992633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27992672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27992629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27992670 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07430364 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02386396 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110209 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07430370 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07430376 - -
          Whole Mouse Genome G4122A A_51_P237512 BC119167.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237512 BC060088.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237512 BC096627.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237512 BC117001.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237512 AK162628.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237512 BC119167.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237512 BC060088.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237512 BC096627.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237512 BC117001.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237512 AK162628.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137597 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07430370 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110209 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09481071 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09157292 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09157294 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09318646 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09157288 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09481064 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09157291 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27992598 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27992600 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27992601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27992598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09605922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09496051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27992625 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10379991 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379990 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379993 - -
          Affymetrix Mouse Gene 1.0 ST Array 10350591 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379994 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379992 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379995 - -
          Affymetrix HT MG-430 PM Array Plate 1437480_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423440_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10350590 - -
          Affymetrix Mouse Gene 1.1 ST Array 10379989 - -
          Mouse Genome 430 2.0 Array 1447413_PM_at - -
          Mouse Genome 430 2.0 Array 1423440_PM_at - -
          Mouse Genome 430 2.0 Array 1458130_PM_at - -
          Mouse Genome 430 2.0 Array 1423440_at - -
          Mouse Genome 430 2.0 Array 1437480_PM_at - -
          Mouse Genome 430 2.0 Array 1447413_at - -
          Mouse Genome 430 2.0 Array 1458130_at - -
          Mouse Genome 430 2.0 Array 1437480_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66140_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66140_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100012393.mm.1 NM_025377.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012393.mm.1 NM_025377.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012393.mm.1 TC1100001329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100012395.mm.1 NM_025377.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012395.mm.1 NM_025377.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012395.mm.1 TC1100001329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100012394.mm.1 NM_025377.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012394.mm.1 NM_025377.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012394.mm.1 TC1100001329.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100012396.mm.1 NM_025377.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012396.mm.1 NM_025377.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100012396.mm.1 TC1100001329.mm.1 MTA TC
          Mouse Expression Set 430 1423440_PM_at - -
          Mouse Expression Set 430 1437480_at - -
          Mouse Expression Set 430 1437480_PM_at - -
          Mouse Expression Set 430 1423440_at - -
          Murine Genome U74v2 Set 160987_r_at - -
          Murine Genome U74v2 Set 160987_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1437480_at - -
          Mouse Genome 430A 2.0 Array 1437480_PM_at - -
          Mouse Genome 430A 2.0 Array 1423440_at - -
          Mouse Genome 430A 2.0 Array 1423440_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1437480_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1437480_at - -
          Affymetrix HT Mouse Genome 430A Array 1423440_at - -
          Affymetrix HT Mouse Genome 430A Array 1423440_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 309759 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6510735 - -
          Illumina mouse-6 v1.0 expression beadchip 540164 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006510735 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000540164 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2665957 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2645490 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2665957 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247197 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2645490 - -
          mouse-6 v1.1 (Illumina) scl0066140.1_151-S - -
          mouse-6 v1.1 (Illumina) scl41091.4_132-S - -
          Illumina Sentrix mouse-6.v1 bead array 540164 - -
          Illumina Sentrix mouse-6.v1 bead array 6510735 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0066140.1_151-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41091.4_132-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540164 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6510735 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2645490 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2789047 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2645490 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2789047 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl41091.4_132-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline