Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02015493_s1
          See other GM6787 GE Assays ›
          Gene Symbol:
          Gm6787
          Gene Name:
          proteasome (prosome, macropain) subunit, beta type 7 pseudogene
          Gene Aliases:
          EG627782
          Chromosome Location:
          Chr.X: 8097087 - 8106701 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02015493_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm6787
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003632.2 4 931 129
          GenBank mRNA AI465359.1 - 1 128

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream Wiskott-Aldrich syndrome homolog (human) Was U42471,
          Wasp
          -15.1kb Anti-sense Chr. X - 8081466 - 8090491 on NCBI Build GRCm38
          Downstream WD repeat domain 13 Wdr13 1700060B08Rik,
          5730411P10Rik,
          DXHXS7467e,
          W51679,
          mMg21
          16.6kb Anti-sense Chr. X - 8123301 - 8132858 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P759266 NR_003632.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P759266 NR_003632.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 166161 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 241559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471700 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 249535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127594 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P759266 NR_003632.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P759266 NR_003632.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P759266 NR_003632.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P759266 NR_003632.2 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P32241757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32241762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14737491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14781883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32241759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32241756 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10598503 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598502 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598504 - -
          Affymetrix HT MG-430 PM Array Plate 1419894_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1449657_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10598501 - -
          Mouse Genome 430 2.0 Array 1419894_at - -
          Mouse Genome 430 2.0 Array 1419894_PM_at - -
          Mouse Genome 430 2.0 Array 1449657_at - -
          Mouse Genome 430 2.0 Array 1449657_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 627782_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 627782_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00000634.mm.1 NR_003632.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000634.mm.1 TC0X00000083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00000633.mm.1 NR_003632.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000633.mm.1 TC0X00000083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00000638.mm.1 NR_003632.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000638.mm.1 TC0X00000083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00000635.mm.1 NR_003632.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000635.mm.1 TC0X00000083.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00000637.mm.1 NR_003632.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000637.mm.1 TC0X00000083.mm.1 MTA TC
          Mouse Expression Set 430 1449657_PM_at - -
          Mouse Expression Set 430 1419894_at - -
          Mouse Expression Set 430 1449657_at - -
          Mouse Expression Set 430 1419894_PM_at - -
          Mouse Genome 430A 2.0 Array 1449657_at - -
          Mouse Genome 430A 2.0 Array 1419894_at - -
          Mouse Genome 430A 2.0 Array 1419894_PM_at - -
          Mouse Genome 430A 2.0 Array 1449657_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419894_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419894_at - -
          Affymetrix HT Mouse Genome 430A Array 1449657_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449657_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160222 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160222 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2864179 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline