Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02026122_g1
          See other GPI1 GE Assays ›
          Gene Symbol:
          Gpi1
          Gene Name:
          glucose phosphate isomerase 1
          Gene Aliases:
          Amf, Gpi, Gpi-1, Gpi-1r, Gpi-1s, Gpi-1t, Gpi1-r, Gpi1-s, Gpi1-t, Gpi1s, MF, NK, NK/GPI, Nlk, Org, Pgi, Phi
          Chromosome Location:
          Chr.7: 34201327 - 34230336 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02026122_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpi1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008155.4 NP_032181.2 13 1251 108
          GenBank mRNA AK031081.1 - 13 1156 108
          AK076424.1 - 11 907 108
          AK133827.1 - 13 1173 108
          AK134890.1 - 13 1185 108
          AK137805.1 - 13 1179 108
          AK147124.1 - 13 1214 108
          AK150341.1 - 13 1182 108
          AK152508.1 - 8 555 108
          AK159838.1 - 13 1155 108
          AK169681.1 - 13 1180 108
          BC086640.1 - 13 1198 6832250 108
          BC088995.1 - 13 1158 6403790 108
          L09104.1 - 4 2390 108
          M14220.1 - 13 1180 108

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpi1

          Panther Classification:

          Molecular Function -

          isomerase

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          in utero embryonic development
          mesoderm formation
          carbohydrate metabolic process
          gluconeogenesis
          glycolytic process
          methylglyoxal biosynthetic process
          erythrocyte homeostasis
          glucose homeostasis
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of neuron apoptotic process
          aldehyde catabolic process
          glucose 6-phosphate metabolic process
          glycolytic process through glucose-6-phosphate
          glucose-6-phosphate isomerase activity
          cytokine activity
          protein binding
          growth factor activity
          isomerase activity
          intramolecular transferase activity
          monosaccharide binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700025281.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025281.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025283.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025283.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025284.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025284.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025291.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025291.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025292.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025292.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025302.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025302.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025285.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025285.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025286.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025286.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025293.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025293.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025289.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025289.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025290.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025290.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025298.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025298.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025300.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025300.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025304.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025304.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025282.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025282.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025295.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025295.mm.1 TC0700002828.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025308.mm.1 NM_008155.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025308.mm.1 TC0700002828.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 900795 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline