Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02028167_s1
          See other GEMIN4 GE Assays ›
          Gene Symbol:
          Gemin4 Fam57a
          Gene Name:
          gem (nuclear organelle) associated protein 4
          family with sequence similarity 57, member A
          Gene Aliases:
          4932415L08Rik, AU015359
          2310047D13Rik, 4932415L08Rik, 5430402E13Rik, 5430420K21Rik, Wdt3
          Chromosome Location:
          Chr.11: 76210571 - 76217572 on Build GRCm38
          Chromosome Location:
          Chr.11: 76179676 - 76209417 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm02028167_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gemin4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177367.3 NP_796341.2 2 2054 149
          GenBank mRNA AK133821.1 - 2 2147 149
          AK141078.1 - 2 2033 149
          AK154061.1 - 2 2035 149
          AK165864.1 - 2 1998 149
          BC031425.1 - 2 1994 3982927 149
          BC062157.1 - 2 2002 6508223 149
          Gene Symbol: Fam57a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK030014.1 - 149
          AK158967.1 - 149

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK158967.1 transcripts.
          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gemin4
          Click here to find siRNAs for Gene Fam57a

          Gene Ontology Categories:

          Function(s) Process(es)

          spliceosomal snRNP assembly
          ribonucleoprotein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27930667 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27930668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27930670 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27930669 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966573 BC031425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966573 BC062157.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966573 AK154061.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966573 AK165864.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966573 AK133821.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P476699 BC031425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P476699 BC062157.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P476699 AK154061.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P476699 AK165864.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P476699 AK133821.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P476699 BC031425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P476699 BC062157.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P476699 AK154061.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P476699 AK165864.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P476699 AK133821.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 743173 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 417008 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837886 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65486 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166279 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P476699 BC031425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P476699 BC062157.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P476699 AK154061.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P476699 AK165864.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P476699 AK133821.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P476699 BC031425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P476699 BC062157.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P476699 AK154061.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P476699 AK165864.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P476699 AK133821.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966573 BC031425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966573 BC062157.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966573 AK154061.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966573 AK165864.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966573 AK133821.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27930652 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166279 - -
          Whole Mouse Genome G4122A A_52_P476699 BC031425.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P476699 BC062157.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P476699 AK154061.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P476699 AK165864.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P476699 AK133821.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P476699 BC031425.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P476699 BC062157.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P476699 AK154061.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P476699 AK165864.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P476699 AK133821.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134694 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166279 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09077671 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09077661 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09077660 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27930668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27930669 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27930670 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27930667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27930667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09022042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09275373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09077661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09672529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09077660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27930614 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10388518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378764 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388519 - -
          Affymetrix HT MG-430 PM Array Plate 1433622_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10378763 - -
          Affymetrix Mouse Gene 1.1 ST Array 10388517 - -
          Mouse Genome 430 2.0 Array 1433622_at - -
          Mouse Genome 430 2.0 Array 1433622_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 276919_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 276919_at - -
          Affymetrix Mouse Clariom S Array PSR1100029831.mm.1 NM_177367.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029831.mm.1 NM_177367.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029831.mm.1 TC1100004294.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100029824.mm.1 NM_177367.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029824.mm.1 NM_177367.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029824.mm.1 TC1100004294.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100029828.mm.1 NM_177367.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029828.mm.1 NM_177367.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029828.mm.1 TC1100004294.mm.1 MTA TC
          Mouse Expression Set 430 1433622_PM_at - -
          Mouse Expression Set 430 1433622_at - -
          Murine Genome U74v2 Set 134711_at - -
          Murine Genome U74v2 Set 134711_PM_at - -
          Murine Genome U74v2 Set 165425_f_at - -
          Murine Genome U74v2 Set 165425_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1433622_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433622_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 687393 - -
          ABI Mouse Genome Survey Microarray 899001 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 130278 - -
          Illumina mouse-6 v1.0 expression beadchip 130278 - -
          Illumina mouse-6 v1.0 expression beadchip 2230075 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002230075 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000130278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223348 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2682362 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2682362 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223348 - -
          mouse-6 v1.1 (Illumina) scl00276919.1_37-S - -
          mouse-6 v1.1 (Illumina) scl00276919.2_158-S - -
          Illumina Sentrix mouse-6.v1 bead array 130278 - -
          Illumina Sentrix mouse-6.v1 bead array 2230075 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00276919.1_37-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00276919.2_158-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 130278 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2230075 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2682362 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1223348 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2963974 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223348 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2682362 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2963974 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00276919.2_158-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline