| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300008563.mm.1 |
NM_153523.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300008563.mm.1 |
NM_153523.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300008563.mm.1 |
TC1300001367.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300008561.mm.1 |
NM_153523.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300008561.mm.1 |
NM_153523.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300008561.mm.1 |
TC1300001367.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300008562.mm.1 |
NM_153523.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300008562.mm.1 |
NM_153523.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300008562.mm.1 |
TC1300001367.mm.1 |
MTA TC |