Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02344423_s1
          See other OLFR1257 GE Assays ›
          Gene Symbol:
          Olfr1257
          Gene Name:
          olfactory receptor 1257
          Gene Aliases:
          MOR232-1
          Chromosome Location:
          Chr.2: 89880828 - 89881757 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02344423_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1257
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146982.1 NP_667193.1 1 262 88
          GenBank mRNA BC137779.1 - 1 296 8860798 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1257

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1987269 NM_146982.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1987269 BC137779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1987269 NM_146982.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1987269 BC137779.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415079 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04600312 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136307 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01396650 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01396641 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01396643 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01396648 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01396637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01828667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01396645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01197873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01396650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415091 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473625 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473624 - -
          Affymetrix Mouse Clariom S Array PSR0200010132.mm.1 TC0200001337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010130.mm.1 TC0200001337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010131.mm.1 NM_146982.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010131.mm.1 NM_146982.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010131.mm.1 TC0200001337.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160426 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160426 - -
          mouse-6 v1.1 (Illumina) GI_49227473-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_2806803 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2806803 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline