Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02599906_s1
          See other GM534 GE Assays ›
          Gene Symbol:
          Gm534
          Gene Name:
          predicted gene 534
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02599906_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm534
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK052085.1 - 1 957 111

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017565 AK052085.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106275 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017565 AK052085.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1164127 AK052085.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11815090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11336070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11800147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29590613 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 490218 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106220451 - -
          Illumina mouse-6 v1.0 expression beadchip 106220451 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257426 - -
          Illumina Sentrix mouse-6.v1 bead array 106220451 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106220451 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257426 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline