Search
Search

Rbm44| Gene Symbol: Rbm44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033408.4 | NP_001028580.1 | 9 - 10 | 2295 | 142 | |
| GenBank mRNA | AK133355.1 | - | 9 - 10 | 2296 | 142 | |
| BC150950.1 | - | 9 - 10 | 2257 | 9087861 | 142 | |
Set Membership: |
|