Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02601633_g1
          See other RPL19 GE Assays ›
          Gene Symbol:
          Rpl19
          Gene Name:
          ribosomal protein L19
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 98023080 - 98030493 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02601633_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001159483.1 NP_001152955.1 5 - 6 517 69
          NM_009078.2 NP_033104.2 5 - 6 531 69
          GenBank mRNA AK010440.1 - 5 - 6 498 69
          AK075817.1 - 5 - 6 499 69
          AK168232.1 - 4 - 5 456 69
          AK168481.1 - 5 - 6 502 69
          AV467392.1 - 2 - 3 159 69
          BC010710.1 - 4 - 5 476 2648593 69
          BC083131.1 - 4 - 5 478 6442192 69
          BC087961.1 - 5 - 6 497 6761831 69
          BC089549.1 - 5 - 6 498 6703351 69
          BC092539.1 - 5 - 6 487 6594084 69
          BU524941.1 - 5 - 6 533 69
          CB203522.1 - 5 - 6 517 69
          M62952.1 - 4 - 5 469 69

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl19

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 676993 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838838 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120519 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152245 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P547021 BC087961.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P547021 BC089549.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P547021 BC092539.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P547021 BU524941.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P547021 BC010710.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P415162 BC087961.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P415162 BC089549.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P415162 BC092539.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P415162 BU524941.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P415162 BC010710.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28054032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28054023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28054034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28054035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28054030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28054026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28054037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28054028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28054038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28054022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28054021 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120519 - -
          Whole Mouse Genome G4122A A_51_P415162 BC087961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P415162 BC089549.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P415162 BC092539.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P415162 BU524941.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P415162 BC010710.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547021 BC087961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547021 BC089549.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547021 BC092539.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547021 BU524941.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P547021 BC010710.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118827 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119186 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120519 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09735965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28054000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09248112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09233848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09635712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09735963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09533770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28054017 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10380834 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570626 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380836 - -
          Affymetrix Mouse Gene 1.0 ST Array 10417666 - -
          Affymetrix Mouse Gene 1.0 ST Array 10556572 - -
          Affymetrix Mouse Gene 1.0 ST Array 10454297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10347918 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380835 - -
          Affymetrix Mouse Gene 1.0 ST Array 10417665 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380837 - -
          Affymetrix Mouse Gene 1.0 ST Array 10422853 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380838 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380839 - -
          Affymetrix HT MG-430 PM Array Plate 1416219_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10380833 - -
          Affymetrix Mouse Gene 1.1 ST Array 10570625 - -
          Affymetrix Mouse Gene 1.1 ST Array 10556571 - -
          Affymetrix Mouse Gene 1.1 ST Array 10422852 - -
          Affymetrix Mouse Gene 1.1 ST Array 10347917 - -
          Affymetrix Mouse Gene 1.1 ST Array 10417664 - -
          Affymetrix Mouse Gene 1.1 ST Array 10454296 - -
          Mouse Genome 430 2.0 Array 1416219_PM_at - -
          Mouse Genome 430 2.0 Array 1445575_at - -
          Mouse Genome 430 2.0 Array 1445575_PM_at - -
          Mouse Genome 430 2.0 Array 1416219_at - -
          Affymetrix Mouse Clariom S Array PSR1100014350.mm.1 NM_001159483.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014350.mm.1 NM_001159483.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014350.mm.1 NM_009078.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014350.mm.1 NM_009078.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014350.mm.1 TC1100004274.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100014351.mm.1 NM_001159483.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014351.mm.1 NM_001159483.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014351.mm.1 NM_009078.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014351.mm.1 NM_009078.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014351.mm.1 TC1100004274.mm.1 MTA TC
          Mouse Expression Set 430 1416219_PM_at - -
          Mouse Expression Set 430 1416219_at - -
          Murine Genome U74v2 Set 97483_PM_at - -
          Murine Genome U74v2 Set 97483_at - -
          Mouse Genome 430A 2.0 Array 1416219_PM_at - -
          Mouse Genome 430A 2.0 Array 1416219_at - -
          Affymetrix HT Mouse Genome 430A Array 1416219_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416219_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5550592 - -
          Illumina mouse-6 v1.0 expression beadchip 5550592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550592 - -
          mouse-6 v1.1 (Illumina) scl019921.5_131-S - -
          Illumina Sentrix mouse-6.v1 bead array 5550592 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl019921.5_131-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550592 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2869225 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2869225 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226762 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl019921.5_131-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline