Search
Search

| Gene Symbol: Dcun1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205361.1 | NP_001192290.1 | 3 - 4 | 569 | 103 | |
| NM_001205362.1 | NP_001192291.1 | 3 - 4 | 529 | 103 | ||
| NM_033623.6 | NP_296372.3 | 3 - 4 | 516 | 103 | ||
| GenBank mRNA | AF198092.1 | - | 3 - 4 | 407 | 103 | |
| AJ305034.1 | - | 3 - 4 | 569 | 103 | ||
| AK008657.1 | - | 3 - 4 | 406 | 103 | ||
| AK139845.1 | - | 3 - 4 | 519 | 103 | ||
| AK155081.1 | - | 3 - 4 | 440 | 103 | ||
| AK165651.1 | - | 3 - 4 | 442 | 103 | ||
| AK165675.1 | - | 3 - 4 | 414 | 103 | ||
| BC020161.1 | - | 3 - 4 | 492 | 3988680 | 103 | |
| BC031666.1 | - | 3 - 4 | 425 | 4012394 | 103 | |
| BC037431.1 | - | 3 - 4 | 425 | 2648141 | 103 | |
| Gene Symbol: Tes3-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029314.1 | - | 1 | 722 | 103 | |
Set Membership: |
|