Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02763224_gH
          See other GM4983 GE Assays ›
          Gene Symbol:
          Gm4983
          Gene Name:
          predicted gene 4983
          Gene Aliases:
          EG245297
          Chromosome Location:
          Chr.12: 17992836 - 18062261 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02763224_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm4983
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC030401.1 - 1948 5362444 94

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07558126 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07558348 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07558436 - -
          Whole Mouse Genome G4122A A_52_P426062 - -
          Whole Mouse Genome G4122A A_52_P74637 BC030401.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07558348 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07558126 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07558436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32977041 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10394838 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394795 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394796 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394797 - -
          Affymetrix Mouse Gene 1.1 ST Array 10394794 - -
          Mouse Genome 430 2.0 Array 1439113_at - -
          Mouse Genome 430 2.0 Array 1439113_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200001291.mm.1 TC1200000152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001297.mm.1 TC1200000152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001295.mm.1 TC1200000152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001294.mm.1 TC1200000152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001298.mm.1 TC1200000152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001293.mm.1 TC1200000152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001292.mm.1 TC1200000152.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001296.mm.1 TC1200000152.mm.1 MTA TC
          Mouse Expression Set 430 1439113_at - -
          Mouse Expression Set 430 1439113_PM_at - -
          Murine Genome U74v2 Set 98344_PM_f_at - -
          Murine Genome U74v2 Set 98344_f_at - -
          Murine Genome U74v2 Set 161128_PM_r_at - -
          Murine Genome U74v2 Set 161128_r_at - -
          Murine Genome U74v2 Set 104641_f_at - -
          Murine Genome U74v2 Set 104641_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1439113_PM_at - -
          Mouse Genome 430A 2.0 Array 1439113_at - -
          Array Name Probe IDs Targets Target Type
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3076680 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3155626 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline